Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-NH-PEG8-Boc | 648.74 | 50 MG
SDP

Catalog No. 5000351816
Encompass_Preferred

Mal-NH-PEG8-Boc is a laboratory chemical used for the manufacture of substances and is intended for research use only.

  • Stable under recommended storage conditions
  • Recommended storage at 4°C, stored under nitrogen
  • Ships at room temperature for less than 2 weeks
  • Appearance: oil

Catalog No. 50-003-51816 Supplier Medchemexpress LLC Supplier No. HY13844750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-NH-PEG8-Boc is a laboratory chemical used for the manufacture of substances and is intended for research use only.

  • Stable under recommended storage conditions
  • Recommended storage at 4°C, stored under nitrogen
  • Ships at room temperature for less than 2 weeks
  • Appearance: oil

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.