Learn More
Medchemexpress LLC Mal-PEG-mal (MW 2000) | 98.4% | 100 MG
SDP
Catalog No. 5000353128
Encompass_Preferred
Mal-PEG-mal (MW 2000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. These compounds contain two different ligands connected by a linker; one targets an E3 ubiquitin ligase, and the other targets a specific protein. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Ligands target E3 ubiquitin ligase and target protein
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-53128 Supplier Medchemexpress LLC Supplier No. HY140719100MG
Mal-PEG-mal (MW 2000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. These compounds contain two different ligands connected by a linker; one targets an E3 ubiquitin ligase, and the other targets a specific protein. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Ligands target E3 ubiquitin ligase and target protein
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins