Learn More
Medchemexpress LLC Mal-PEG-mal (MW 2000) | 98.40% | 250 MG
SDP
Catalog No. 5000352844
Encompass_Preferred
Mal-PEG-mal (MW 2000) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These PROTAC molecules incorporate two distinct ligands connected by a linker: one targets an E3 ubiquitin ligase, while the other targets the desired protein. This mechanism enables PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Purity of 98.40%
  • Solid appearance
Catalog No. 50-003-52844 Supplier Medchemexpress LLC Supplier No. 250MGHY140719
Mal-PEG-mal (MW 2000) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These PROTAC molecules incorporate two distinct ligands connected by a linker: one targets an E3 ubiquitin ligase, while the other targets the desired protein. This mechanism enables PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Purity of 98.40%
  • Solid appearance