Learn More
Medchemexpress LLC Mal-PEG-mal (MW 5000) | 95.0% | 250 MG
SDP
Catalog No. 5000352716
Encompass_Preferred
Mal-PEG-mal (MW 5000) is a PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs, which are molecules designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. PROTACs function by connecting a ligand for an E3 ubiquitin ligase with a ligand for a target protein via a linker.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52716 Supplier Medchemexpress LLC Supplier No. 250MGHY140721
Mal-PEG-mal (MW 5000) is a PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs, which are molecules designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. PROTACs function by connecting a ligand for an E3 ubiquitin ligase with a ligand for a target protein via a linker.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins