Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-PEG1-NHS ester | 1807518-72-4 | MFCD26127783 | 99.6% | 310.26 g/mol | C13H14N2O7 | 250 MG
SDP

Catalog No. 5000203487
Encompass_Preferred

Mal-PEG1-NHS ester is a PEG-based bifunctional linker containing a maleimide group and an N-hydroxysuccinimide (NHS) ester. It is used as a non-cleavable linker for antibody-drug conjugates and as a PEG linker in PROTAC synthesis to connect thiol- and amine-containing molecules. For research use only.

  • Contains maleimide and NHS ester reactive groups
  • PEG1 spacer increases solubility and flexibility
  • Molecular weight 310.26 g/mol; formula C13H14N2O7
  • High purity (≈99.6%) suitable for conjugation reactions
  • Available in small to multi-gram pack sizes, including 250 mg
  • Store powder at -20°C; solvent solutions stable at -80°C (6 months)

Catalog No. 50-002-03487 Supplier Medchemexpress LLC Supplier No. HY126886250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-PEG1-NHS ester is a PEG-based bifunctional linker containing a maleimide group and an N-hydroxysuccinimide (NHS) ester. It is used as a non-cleavable linker for antibody-drug conjugates and as a PEG linker in PROTAC synthesis to connect thiol- and amine-containing molecules. For research use only.

  • Contains maleimide and NHS ester reactive groups
  • PEG1 spacer increases solubility and flexibility
  • Molecular weight 310.26 g/mol; formula C13H14N2O7
  • High purity (≈99.6%) suitable for conjugation reactions
  • Available in small to multi-gram pack sizes, including 250 mg
  • Store powder at -20°C; solvent solutions stable at -80°C (6 months)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.