Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-PEG1-NHS ester | 1807518-72-4 | MFCD26127783 | 99.6% | 310.26 g/mol | C13H14N2O7 | 5 G
SDP

Catalog No. 5000203158
Encompass_Preferred

Mal-PEG1-NHS ester is a PEG-based, heterobifunctional linker supplied for research use that functions as a non-cleavable ADC linker and as a PEG-based PROTAC linker in small-molecule and bioconjugation synthesis.

  • used for antibody-drug conjugate and PROTAC synthesis
  • high purity suitable for research applications
  • white to off-white solid appearance
  • molecular weight 310.26 g/mol and formula C13H14N2O7
  • storage: powder at -20 °C (stable for 3 years)
  • storage in solvent: -80 °C (6 months) or -20 °C (1 month)
  • packaging sizes include 100 mg, 250 mg, 1 g, 5 g, 10 g, 50 g

Catalog No. 50-002-03158 Supplier Medchemexpress LLC Supplier No. HY1268865G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-PEG1-NHS ester is a PEG-based, heterobifunctional linker supplied for research use that functions as a non-cleavable ADC linker and as a PEG-based PROTAC linker in small-molecule and bioconjugation synthesis.

  • used for antibody-drug conjugate and PROTAC synthesis
  • high purity suitable for research applications
  • white to off-white solid appearance
  • molecular weight 310.26 g/mol and formula C13H14N2O7
  • storage: powder at -20 °C (stable for 3 years)
  • storage in solvent: -80 °C (6 months) or -20 °C (1 month)
  • packaging sizes include 100 mg, 250 mg, 1 g, 5 g, 10 g, 50 g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.