Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-PEG10-NHS ester | 3055858-73-3 | 98.0% | 706.73 | 50 MG
SDP

Catalog No. 5000352001
Encompass_Preferred

Mal-PEG10-NHS ester is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker, with one ligand for an E3 ubiquitin ligase and the other for the target protein, functioning by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Selectively degrades target proteins through the ubiquitin-proteasome system
  • High purity: 98.0%
  • Molecular weight: 706.73
  • Appearance: colorless to light yellow oil
  • Solubility: 100 mg/mL in DMSO

Catalog No. 50-003-52001 Supplier Medchemexpress LLC Supplier No. HY13872650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-PEG10-NHS ester is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker, with one ligand for an E3 ubiquitin ligase and the other for the target protein, functioning by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Selectively degrades target proteins through the ubiquitin-proteasome system
  • High purity: 98.0%
  • Molecular weight: 706.73
  • Appearance: colorless to light yellow oil
  • Solubility: 100 mg/mL in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.