Learn More
Broadpharm Mal-PEG12-NHS ester, 100mg
Catalog No. NC1666659
Encompass
Formula: C35H58N2O18, Purity: 96%, MW: 794.9, Solubility: DMSO, DCM, DMF
Catalog No. NC1666659 Supplier Broadpharm Supplier No. BP24079100MG
Formula: C35H58N2O18, Purity: 96%, MW: 794.9, Solubility: DMSO, DCM, DMF