Learn More
Medchemexpress LLC Mal-PEG12-NHS ester | 2669737-09-9 | MFCD31811553 | 97.0% | C35H58N2O18 | 10MG
SDP
Catalog No. 5000259756
Encompass_Preferred
Mal-PEG12-NHS ester is a heterobifunctional PEG12 linker bearing a maleimide and an N-hydroxysuccinimide (NHS) ester. It enables efficient, site-selective conjugation of sulfhydryl-containing molecules to primary amines through a hydrophilic PEG12 spacer, and is commonly used in PROTAC synthesis and general bioconjugation workflows. Molecular weight ≈794.84 g·mol⁻¹; CAS 2669737-09-9; typical purity ≈97%.
  • Heterobifunctional maleimide-NHS chemistry for thiol-to-amine conjugation.
  • PEG12 spacer increases solubility and reduces steric hindrance.
  • Suitable for PROTAC linker synthesis and general bioconjugation.
  • High purity (~97%) for reliable analytical and preparative use.
  • Available in small-scale quantities for research applications.
Catalog No. 50-002-59756 Supplier Medchemexpress LLC Supplier No. HY14096310MG
Mal-PEG12-NHS ester is a heterobifunctional PEG12 linker bearing a maleimide and an N-hydroxysuccinimide (NHS) ester. It enables efficient, site-selective conjugation of sulfhydryl-containing molecules to primary amines through a hydrophilic PEG12 spacer, and is commonly used in PROTAC synthesis and general bioconjugation workflows. Molecular weight ≈794.84 g·mol⁻¹; CAS 2669737-09-9; typical purity ≈97%.
  • Heterobifunctional maleimide-NHS chemistry for thiol-to-amine conjugation.
  • PEG12 spacer increases solubility and reduces steric hindrance.
  • Suitable for PROTAC linker synthesis and general bioconjugation.
  • High purity (~97%) for reliable analytical and preparative use.
  • Available in small-scale quantities for research applications.