Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-PEG2-gly-gly-phe-gly-exatecan | 1599439-54-9 | 98.6% | 1149.18 | C57H65FN10O15 | 5 MG
SDP

Catalog No. 5000204310
Encompass_Preferred

A drug-linker conjugate used in antibody-drug conjugate (ADC) synthesis and cancer research. It consists of an exatecan payload linked via a maleimide-PEG2-gly-gly-phe-gly linker. For research use only.

  • Suitable for antibody-drug conjugate synthesis
  • High purity: 98.6%
  • Molecular weight: 1149.18 g/mol
  • Chemical formula: C57H65FN10O15
  • Soluble in DMSO
  • Store at -20°C and protect from light

Catalog No. 50-002-04310 Supplier Medchemexpress LLC Supplier No. HY1537955MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A drug-linker conjugate used in antibody-drug conjugate (ADC) synthesis and cancer research. It consists of an exatecan payload linked via a maleimide-PEG2-gly-gly-phe-gly linker. For research use only.

  • Suitable for antibody-drug conjugate synthesis
  • High purity: 98.6%
  • Molecular weight: 1149.18 g/mol
  • Chemical formula: C57H65FN10O15
  • Soluble in DMSO
  • Store at -20°C and protect from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.