Learn More
Medchemexpress LLC Mal-PEG2-NHS | 329364-72-9 | MFCD34471337 | 95.0% | C14H16N2O8 | 10MG
SDP
Catalog No. 5000260558
Encompass_Preferred
Mal-PEG2-NHS is a PEG-based bifunctional linker bearing a maleimide and an N-hydroxysuccinimide (NHS) ester. It enables site-selective conjugation by reacting the maleimide with thiols and the NHS ester with primary amines, facilitating assembly of bifunctional molecules such as PROTACs and other bioconjugates.
  • Contains maleimide and NHS ester functional groups.
  • Provides a two-unit PEG spacer for flexibility and solubility.
  • Enables thiol- and amine-directed conjugation chemistries.
  • Suitable for PROTAC synthesis and general bioconjugation applications.
  • High purity (~95%) for reliable synthetic performance.
Catalog No. 50-002-60558 Supplier Medchemexpress LLC Supplier No. HY13843010MG
Mal-PEG2-NHS is a PEG-based bifunctional linker bearing a maleimide and an N-hydroxysuccinimide (NHS) ester. It enables site-selective conjugation by reacting the maleimide with thiols and the NHS ester with primary amines, facilitating assembly of bifunctional molecules such as PROTACs and other bioconjugates.
  • Contains maleimide and NHS ester functional groups.
  • Provides a two-unit PEG spacer for flexibility and solubility.
  • Enables thiol- and amine-directed conjugation chemistries.
  • Suitable for PROTAC synthesis and general bioconjugation applications.
  • High purity (~95%) for reliable synthetic performance.