Learn More
Medchemexpress LLC Mal-PEG2-NHS ester | 1433997-01-3 | 98.6% | 354.31 g/mol | C15H18N2O8 | 1 G
SDP
Catalog No. 5000201890
Encompass_Preferred
Mal-PEG2-NHS ester is a non-cleavable ADC linker combining a maleimide reactive group, a two-unit polyethylene glycol spacer, and an N-hydroxysuccinimide (NHS) ester for amine coupling. It is supplied as a purified reagent for bioconjugation and linker chemistry applications.
  • Non-cleavable maleimide-NHS bifunctional linker
  • PEG2 spacer improves solubility and reduces steric hindrance
  • NHS ester enables efficient coupling to primary amines
  • Maleimide group enables thiol-selective conjugation
  • High purity suitable for ADC synthesis and bioconjugation
Catalog No. 50-002-01890 Supplier Medchemexpress LLC Supplier No. HY1265041G
Mal-PEG2-NHS ester is a non-cleavable ADC linker combining a maleimide reactive group, a two-unit polyethylene glycol spacer, and an N-hydroxysuccinimide (NHS) ester for amine coupling. It is supplied as a purified reagent for bioconjugation and linker chemistry applications.
  • Non-cleavable maleimide-NHS bifunctional linker
  • PEG2 spacer improves solubility and reduces steric hindrance
  • NHS ester enables efficient coupling to primary amines
  • Maleimide group enables thiol-selective conjugation
  • High purity suitable for ADC synthesis and bioconjugation