Learn More
Medchemexpress LLC Mal-PEG2-NHS ester | 1433997-01-3 | MFCD24539465 | 98.6% | 354.31 | C15H18N2O8 | 100 MG
SDP
Catalog No. 5000202961
Encompass_Preferred
Mal-PEG2-NHS ester is a bifunctional PEG linker designed for bioconjugation, combining a maleimide moiety for thiol coupling with an NHS ester for amine coupling to enable antibody-drug conjugate (ADC) and other conjugation chemistries.
  • Non-cleavable dual-reactive linker enabling site-specific conjugation.
  • Two-unit PEG spacer increases hydrophilicity and solubility.
  • Maleimide group reacts selectively with thiol groups.
  • NHS ester reacts with primary amines for stable amide bond formation.
  • Available in small-scale quantities suitable for research use.
  • Stored and handled to preserve stability: long-term frozen storage recommended.
Catalog No. 50-002-02961 Supplier Medchemexpress LLC Supplier No. HY126504100MG
Mal-PEG2-NHS ester is a bifunctional PEG linker designed for bioconjugation, combining a maleimide moiety for thiol coupling with an NHS ester for amine coupling to enable antibody-drug conjugate (ADC) and other conjugation chemistries.
  • Non-cleavable dual-reactive linker enabling site-specific conjugation.
  • Two-unit PEG spacer increases hydrophilicity and solubility.
  • Maleimide group reacts selectively with thiol groups.
  • NHS ester reacts with primary amines for stable amide bond formation.
  • Available in small-scale quantities suitable for research use.
  • Stored and handled to preserve stability: long-term frozen storage recommended.