Learn More
Medchemexpress LLC Mal-PEG2-NHS ester | 1433997-01-3 | MFCD24539465 | 98.6% | 354.31 g/mol | C15H18N2O8 | 250 MG
SDP
Catalog No. 5000203971
Encompass_Preferred
Mal-PEG2-NHS ester is a noncleavable antibody-drug conjugate (ADC) linker containing a maleimide group for thiol coupling, a two-unit polyethylene glycol (PEG2) spacer, and an N-hydroxysuccinimide (NHS) ester for amine coupling. The compound is supplied for research use and is not for human therapeutic use.
  • Noncleavable ADC linker with maleimide and NHS ester functional groups.
  • PEG2 spacer increases hydrophilicity and reduces aggregation.
  • Facilitates conjugation between thiol-containing and amine-containing molecules.
  • High purity suitable for research applications.
  • Soluble in DMSO; may require ultrasonic assistance for full dissolution.
Catalog No. 50-002-03971 Supplier Medchemexpress LLC Supplier No. HY126504250MG
Mal-PEG2-NHS ester is a noncleavable antibody-drug conjugate (ADC) linker containing a maleimide group for thiol coupling, a two-unit polyethylene glycol (PEG2) spacer, and an N-hydroxysuccinimide (NHS) ester for amine coupling. The compound is supplied for research use and is not for human therapeutic use.
  • Noncleavable ADC linker with maleimide and NHS ester functional groups.
  • PEG2 spacer increases hydrophilicity and reduces aggregation.
  • Facilitates conjugation between thiol-containing and amine-containing molecules.
  • High purity suitable for research applications.
  • Soluble in DMSO; may require ultrasonic assistance for full dissolution.