Learn More
Medchemexpress LLC Mal-PEG2-oxyamine | 1146245-73-9 | 95.0% | C10H16N2O5 | 10MG
SDP
Catalog No. 5000259482
Encompass_Preferred
Mal-PEG2-oxyamine is a bifunctional PEG-based linker for use in the synthesis of PROTACs. It features a maleimide group and an aminooxy (oxyamine) group connected by a short PEG2 spacer, permitting orthogonal conjugation to thiol and carbonyl functionalities for targeted protein degradation research. The product is supplied with a reported purity of 95% and includes storage and solubility guidance for research applications.
  • Bifunctional maleimide and aminooxy reactive groups.
  • PEG2 spacer provides a short, flexible linkage.
  • Enables selective conjugation to thiol and carbonyl partners.
  • Reported purity supports synthetic and analytical use.
  • Includes storage and solubility recommendations for stability.
Catalog No. 50-002-59482 Supplier Medchemexpress LLC Supplier No. HY13345410MG
Mal-PEG2-oxyamine is a bifunctional PEG-based linker for use in the synthesis of PROTACs. It features a maleimide group and an aminooxy (oxyamine) group connected by a short PEG2 spacer, permitting orthogonal conjugation to thiol and carbonyl functionalities for targeted protein degradation research. The product is supplied with a reported purity of 95% and includes storage and solubility guidance for research applications.
  • Bifunctional maleimide and aminooxy reactive groups.
  • PEG2 spacer provides a short, flexible linkage.
  • Enables selective conjugation to thiol and carbonyl partners.
  • Reported purity supports synthetic and analytical use.
  • Includes storage and solubility recommendations for stability.