Learn More
Medchemexpress LLC Mal-PEG2-oxyamine TFA | 00-00-0 | 96.5% | 358.27 g/mol | C12H17F3N2O7 | 5 MG
SDP
Catalog No. 5000262694
Encompass_Preferred
Mal-PEG2-oxyamine TFA is a PEG-based PROTAC linker containing a maleimide and an oxyamine (trifluoroacetate salt) joined by a PEG2 spacer. It is designed for orthogonal bioconjugation-maleimide reacts with thiols and oxyamine forms oximes with aldehydes/ketones-making it suitable for assembling PROTACs and other conjugates. The product is supplied as a colorless to light yellow liquid with high purity and good DMSO solubility.
  • Enables thiol conjugation via maleimide chemistry.
  • Permits oxime ligation through the oxyamine functional group.
  • Short PEG2 spacer provides moderate linker flexibility.
  • High solubility in DMSO (≈250 mg/mL) for convenient stock solutions.
  • High purity (~96.5%) suitable for research synthesis.
  • Stable when stored sealed and protected from moisture.
Catalog No. 50-002-62694 Supplier Medchemexpress LLC Supplier No. HY133454A5MG
Mal-PEG2-oxyamine TFA is a PEG-based PROTAC linker containing a maleimide and an oxyamine (trifluoroacetate salt) joined by a PEG2 spacer. It is designed for orthogonal bioconjugation-maleimide reacts with thiols and oxyamine forms oximes with aldehydes/ketones-making it suitable for assembling PROTACs and other conjugates. The product is supplied as a colorless to light yellow liquid with high purity and good DMSO solubility.
  • Enables thiol conjugation via maleimide chemistry.
  • Permits oxime ligation through the oxyamine functional group.
  • Short PEG2 spacer provides moderate linker flexibility.
  • High solubility in DMSO (≈250 mg/mL) for convenient stock solutions.
  • High purity (~96.5%) suitable for research synthesis.
  • Stable when stored sealed and protected from moisture.