Learn More
Medchemexpress LLC Mal-PEG2-oxyamine TFA | 00-00-0 | MFCD00209801 | 96.5% | C12H17F3N2O7 | 10MG
SDP
Catalog No. 5000261419
Encompass_Preferred
Mal-PEG2-oxyamine TFA is a PEG-based bifunctional linker supplied as the trifluoroacetic acid (TFA) salt for use in PROTAC synthesis and other bioconjugation applications. It combines a maleimide reactive group with an oxyamine functional handle, and a short PEG2 spacer to enhance solubility and flexibility.
  • Bifunctional linker enabling thiol and carbonyl conjugation.
  • Maleimide permits rapid, thiol-selective coupling under mild conditions.
  • Oxyamine enables chemoselective oxime ligation to aldehydes and ketones.
  • PEG2 spacer increases aqueous solubility and reduces steric hindrance.
  • Provided as TFA salt to improve stability and handling.
  • High purity suitable for research and synthetic workflows.
Catalog No. 50-002-61419 Supplier Medchemexpress LLC Supplier No. HY133454A10MG
Mal-PEG2-oxyamine TFA is a PEG-based bifunctional linker supplied as the trifluoroacetic acid (TFA) salt for use in PROTAC synthesis and other bioconjugation applications. It combines a maleimide reactive group with an oxyamine functional handle, and a short PEG2 spacer to enhance solubility and flexibility.
  • Bifunctional linker enabling thiol and carbonyl conjugation.
  • Maleimide permits rapid, thiol-selective coupling under mild conditions.
  • Oxyamine enables chemoselective oxime ligation to aldehydes and ketones.
  • PEG2 spacer increases aqueous solubility and reduces steric hindrance.
  • Provided as TFA salt to improve stability and handling.
  • High purity suitable for research and synthetic workflows.