Learn More
Medchemexpress LLC Mal-PEG2-Val-Cit-PAB-OH | 2055041-38-6 | 99.5% | 618.68 g/mol | C29H42N6O9 | 5 MG
SDP
Catalog No. 5000203831
Encompass_Preferred
Mal-PEG2-Val-Cit-PAB-OH is a cleavable antibody-drug conjugate (ADC) linker featuring a maleimide group, a PEG2 spacer, a valine-citrulline dipeptide cleavable linker, and a para-aminobenzyl (PAB) self-immolative spacer. It is supplied as a solid for research use and is intended for use in ADC and PROTAC synthesis.
  • Cleavable Val-Cit dipeptide for enzyme-triggered payload release.
  • Maleimide functional group enables thiol conjugation to antibodies or payloads.
  • PEG2 spacer improves solubility and flexibility of the linker.
  • PAB self-immolative spacer enables payload release after enzymatic cleavage.
  • High purity (99.49% by COA) and molecular weight ~618.68 g/mol.
Catalog No. 50-002-03831 Supplier Medchemexpress LLC Supplier No. HY1302225MG
Mal-PEG2-Val-Cit-PAB-OH is a cleavable antibody-drug conjugate (ADC) linker featuring a maleimide group, a PEG2 spacer, a valine-citrulline dipeptide cleavable linker, and a para-aminobenzyl (PAB) self-immolative spacer. It is supplied as a solid for research use and is intended for use in ADC and PROTAC synthesis.
  • Cleavable Val-Cit dipeptide for enzyme-triggered payload release.
  • Maleimide functional group enables thiol conjugation to antibodies or payloads.
  • PEG2 spacer improves solubility and flexibility of the linker.
  • PAB self-immolative spacer enables payload release after enzymatic cleavage.
  • High purity (99.49% by COA) and molecular weight ~618.68 g/mol.