Learn More
Medchemexpress LLC Mal-PEG2-VCP-Eribulin | 2130869-18-8 | 99.8% | 1374.57 | 5 MG
SDP
Catalog No. 5000364629
Encompass_Preferred
Mal-PEG2-VCP-Eribulin is a drug-linker conjugate for ADCs, combining an ADCs linker (Mal-PEG2-VCP) with Eribulin. Eribulin is a mechanistically unique microtubule inhibitor that inhibits the proliferation of cancer cells by binding to microtubule proteins and microtubules. This Eribulin-based agent is designed for antibody conjugates and is intended for research use only.
  • Drug-linker conjugate for ADCs
  • Contains ADCs linker (Mal-PEG2-VCP)
  • Incorporates Eribulin, a microtubule inhibitor
  • Inhibits cancer cell proliferation
  • Suitable for antibody conjugates
  • For research use only
Catalog No. 50-003-64629 Supplier Medchemexpress LLC Supplier No. 5MGHY128870
Mal-PEG2-VCP-Eribulin is a drug-linker conjugate for ADCs, combining an ADCs linker (Mal-PEG2-VCP) with Eribulin. Eribulin is a mechanistically unique microtubule inhibitor that inhibits the proliferation of cancer cells by binding to microtubule proteins and microtubules. This Eribulin-based agent is designed for antibody conjugates and is intended for research use only.
  • Drug-linker conjugate for ADCs
  • Contains ADCs linker (Mal-PEG2-VCP)
  • Incorporates Eribulin, a microtubule inhibitor
  • Inhibits cancer cell proliferation
  • Suitable for antibody conjugates
  • For research use only