Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-PEG2-VCP-Eribulin | 2130869-18-8 | MFCD32068377 | >98.0% | 1374.57 | C70H99N7O21 | 1 MG
SDP

Catalog No. 5000203284
Encompass_Preferred

Mal-PEG2-VCP-Eribulin is an Eribulin-based drug-linker conjugate intended for antibody-drug conjugate (ADC) research. It comprises the Mal-PEG2-VCP linker covalently attached to Eribulin, a mechanistically unique microtubule inhibitor, and is supplied in small pack sizes for conjugation and preclinical studies.

  • Contains Mal-PEG2-VCP linker conjugated to Eribulin.
  • Mechanism: microtubule inhibitor for use as a cytotoxic payload.
  • Intended use: building block for antibody-drug conjugates and ADC research.
  • Key identifiers: CAS 2130869-18-8, formula C70H99N7O21, molecular weight ~1374.6.
  • Available in small pack sizes suitable for research (for example, 1 mg).
  • Typical reported purity: greater than 98%.

Catalog No. 50-002-03284 Supplier Medchemexpress LLC Supplier No. HY1288701MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-PEG2-VCP-Eribulin is an Eribulin-based drug-linker conjugate intended for antibody-drug conjugate (ADC) research. It comprises the Mal-PEG2-VCP linker covalently attached to Eribulin, a mechanistically unique microtubule inhibitor, and is supplied in small pack sizes for conjugation and preclinical studies.

  • Contains Mal-PEG2-VCP linker conjugated to Eribulin.
  • Mechanism: microtubule inhibitor for use as a cytotoxic payload.
  • Intended use: building block for antibody-drug conjugates and ADC research.
  • Key identifiers: CAS 2130869-18-8, formula C70H99N7O21, molecular weight ~1374.6.
  • Available in small pack sizes suitable for research (for example, 1 mg).
  • Typical reported purity: greater than 98%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.