Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-PEG2-VCP-eribulin | 2130869-18-8 | MFCD32068377 | 99.8% | 1374.57 | C70H99N7O21 | 10 MG
SDP

Catalog No. 5000204190
Encompass_Preferred

Mal-PEG2-VCP-eribulin is a drug-linker conjugate composed of an ADC linker (Mal-PEG2-VCP) covalently attached to eribulin. It is supplied as a solid for research use in the development of antibody-drug conjugates and related preclinical studies.

  • Consists of an ADC linker conjugated to eribulin.
  • CAS number 2130869-18-8.
  • Molecular weight 1374.57.
  • Chemical formula C70H99N7O21.
  • Purity 99.8%.
  • Supplied as a 10 MG solid; other pack sizes available on request.
  • Store at -20°C, protect from light, under inert atmosphere; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-04190 Supplier Medchemexpress LLC Supplier No. HY12887010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-PEG2-VCP-eribulin is a drug-linker conjugate composed of an ADC linker (Mal-PEG2-VCP) covalently attached to eribulin. It is supplied as a solid for research use in the development of antibody-drug conjugates and related preclinical studies.

  • Consists of an ADC linker conjugated to eribulin.
  • CAS number 2130869-18-8.
  • Molecular weight 1374.57.
  • Chemical formula C70H99N7O21.
  • Purity 99.8%.
  • Supplied as a 10 MG solid; other pack sizes available on request.
  • Store at -20°C, protect from light, under inert atmosphere; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.