Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-PEG2-VCP-NB | 2395887-69-9 | 98.3% | 783.78 g/mol | C36H45N7O13 | 50 MG
SDP

Catalog No. 5000202469
Encompass_Preferred

Mal-PEG2-VCP-NB is a cleavable antibody-drug conjugate (ADC) linker featuring a maleimide reactive group, a two-unit polyethylene glycol (PEG2) spacer, and a VCP NB moiety. It is supplied as a research chemical for ADC conjugation and formulation studies.

  • Cleavable ADC linker containing a maleimide reactive group.
  • Two-unit PEG spacer to improve solubility and flexibility.
  • VCP NB moiety for payload attachment.
  • Purity 98.25% and molecular weight 783.78 g/mol.
  • Soluble in DMSO (100 mg/mL); sonication may be required.
  • Supplied as a 50 mg powder; store at 4°C under nitrogen; in solution store -80°C up to 6 months.

Catalog No. 50-002-02469 Supplier Medchemexpress LLC Supplier No. HY13008450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-PEG2-VCP-NB is a cleavable antibody-drug conjugate (ADC) linker featuring a maleimide reactive group, a two-unit polyethylene glycol (PEG2) spacer, and a VCP NB moiety. It is supplied as a research chemical for ADC conjugation and formulation studies.

  • Cleavable ADC linker containing a maleimide reactive group.
  • Two-unit PEG spacer to improve solubility and flexibility.
  • VCP NB moiety for payload attachment.
  • Purity 98.25% and molecular weight 783.78 g/mol.
  • Soluble in DMSO (100 mg/mL); sonication may be required.
  • Supplied as a 50 mg powder; store at 4°C under nitrogen; in solution store -80°C up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.