Learn More
Medchemexpress LLC Mal-PEG24-NHS ester | 2226733-37-3 | MFCD11041160 | 95.0% | C62H111N3O31 | 10MG
SDP
Catalog No. 5000259848
Encompass_Preferred
Mal-PEG24-NHS ester is a polyethylene glycol (PEG)-based bifunctional linker bearing a maleimide and an N-hydroxysuccinimide (NHS) ester. It enables thiol-to-maleimide and amine-to-NHS conjugation for assembly of PROTACs and other bioconjugates; supplied as a colorless to light-yellow oil with high purity and good aqueous solubility.
  • Enables thiol and amine conjugation via maleimide and NHS ester chemistry.
  • Long PEG24 spacer increases solubility and reduces steric hindrance.
  • High purity supports reliable reaction performance.
  • High aqueous solubility simplifies handling and formulation.
  • Suitable for PROTAC synthesis and general bioconjugation workflows.
Catalog No. 50-002-59848 Supplier Medchemexpress LLC Supplier No. HY13581910MG
Mal-PEG24-NHS ester is a polyethylene glycol (PEG)-based bifunctional linker bearing a maleimide and an N-hydroxysuccinimide (NHS) ester. It enables thiol-to-maleimide and amine-to-NHS conjugation for assembly of PROTACs and other bioconjugates; supplied as a colorless to light-yellow oil with high purity and good aqueous solubility.
  • Enables thiol and amine conjugation via maleimide and NHS ester chemistry.
  • Long PEG24 spacer increases solubility and reduces steric hindrance.
  • High purity supports reliable reaction performance.
  • High aqueous solubility simplifies handling and formulation.
  • Suitable for PROTAC synthesis and general bioconjugation workflows.