Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-PEG3-C1-NHS ester | 2230016-26-7 | 95.0% | 384.34 | C16H20N2O9 | 5 MG
SDP

Catalog No. 5000202371
Encompass_Preferred

Mal-PEG3-C1-NHS ester is a non-cleavable three-unit polyethylene glycol (PEG3) linker bearing a maleimide and an N-hydroxysuccinimide (NHS) ester for conjugation to thiol and amine groups in antibody-drug conjugates. Supplied as a solid with typical purity 95.0%, it is provided with supporting documentation for safe handling and characterization.

  • Non-cleavable three-unit PEG linker for stable conjugation.
  • Contains maleimide and NHS ester functional groups for thiol and amine coupling.
  • Molecular weight 384.34 g/mol.
  • Purity 95.0%.
  • CAS number 2230016-26-7 for chemical identification.
  • Available in 5 mg, 10 mg, 25 mg, and 50 mg sizes; supplied for reconstitution in DMSO at indicated concentrations.

Catalog No. 50-002-02371 Supplier Medchemexpress LLC Supplier No. HY1335815MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-PEG3-C1-NHS ester is a non-cleavable three-unit polyethylene glycol (PEG3) linker bearing a maleimide and an N-hydroxysuccinimide (NHS) ester for conjugation to thiol and amine groups in antibody-drug conjugates. Supplied as a solid with typical purity 95.0%, it is provided with supporting documentation for safe handling and characterization.

  • Non-cleavable three-unit PEG linker for stable conjugation.
  • Contains maleimide and NHS ester functional groups for thiol and amine coupling.
  • Molecular weight 384.34 g/mol.
  • Purity 95.0%.
  • CAS number 2230016-26-7 for chemical identification.
  • Available in 5 mg, 10 mg, 25 mg, and 50 mg sizes; supplied for reconstitution in DMSO at indicated concentrations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.