Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-PEG3-C1-NHS ester | 2230016-26-7 | 95.0% | 384.34 g/mol | C16H20N2O9 | 25 MG
SDP

Catalog No. 5000202374
Encompass_Preferred

Mal-PEG3-C1-NHS ester is a non-cleavable, three-unit polyethylene glycol (PEG) linker bearing maleimide and N-hydroxysuccinimide (NHS) ester functional groups, designed for use in antibody-drug conjugate (ADC) synthesis and related bioconjugation research.

  • Non-cleavable three-unit PEG linker
  • Contains maleimide and NHS ester functional groups
  • Enables conjugation to thiols and amines for ADC assembly
  • Purity ≥95.0% and molecular weight 384.34 g/mol
  • Available in small milligram packaging for research applications

Catalog No. 50-002-02374 Supplier Medchemexpress LLC Supplier No. HY13358125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-PEG3-C1-NHS ester is a non-cleavable, three-unit polyethylene glycol (PEG) linker bearing maleimide and N-hydroxysuccinimide (NHS) ester functional groups, designed for use in antibody-drug conjugate (ADC) synthesis and related bioconjugation research.

  • Non-cleavable three-unit PEG linker
  • Contains maleimide and NHS ester functional groups
  • Enables conjugation to thiols and amines for ADC assembly
  • Purity ≥95.0% and molecular weight 384.34 g/mol
  • Available in small milligram packaging for research applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.