Learn More
Medchemexpress LLC Mal-PEG3-C1-NHS ester | 2230016-26-7 | 95.0% | 384.34 g/mol | C16H20N2O9 | 50 MG
SDP
Catalog No. 5000203673
Encompass_Preferred
Mal-PEG3-C1-NHS ester is a non-cleavable 3-unit polyethylene glycol (PEG) linker bearing a maleimide and an N-hydroxysuccinimide (NHS) ester. It is supplied for research use to facilitate conjugation between amine and thiol groups during antibody-drug conjugate (ADC) synthesis, and is provided as a solution for low-temperature storage.
  • Non-cleavable 3-unit polyethylene glycol linker.
  • Terminal maleimide and NHS ester functional groups for thiol and amine coupling.
  • Suitable for antibody-drug conjugate (ADC) synthesis.
  • Purity approximately 95.0%.
  • Supplied as a solution; store at -20°C for up to 2 years.
Catalog No. 50-002-03673 Supplier Medchemexpress LLC Supplier No. HY13358150MG
Mal-PEG3-C1-NHS ester is a non-cleavable 3-unit polyethylene glycol (PEG) linker bearing a maleimide and an N-hydroxysuccinimide (NHS) ester. It is supplied for research use to facilitate conjugation between amine and thiol groups during antibody-drug conjugate (ADC) synthesis, and is provided as a solution for low-temperature storage.
  • Non-cleavable 3-unit polyethylene glycol linker.
  • Terminal maleimide and NHS ester functional groups for thiol and amine coupling.
  • Suitable for antibody-drug conjugate (ADC) synthesis.
  • Purity approximately 95.0%.
  • Supplied as a solution; store at -20°C for up to 2 years.