Learn More
Carbosynth LLC. Mal-PEG3-Glu[Cyclo(Arg-Gly-Asp-D-Tyr-Lys)]2, 5mg, PEG3 = 11-amino-3,6,9-Trioxaundecanoic acid, MW: 1690.79, Molecular formula: C74H107N21O25
SDP
Supplier:  Carbosynth LLC. RGD3993PI5MG
Encompass_Preferred
Synonyms: Mal-PEG3-Glu[cyclo (Arg-Gly-Asp-D-Tyr-Lys)]2, Mal-PEG3-E[c(RGDwK)]2, Mal-PEG3-E[Cyclo(RGDyK)]2PEG3 = 11-Amino-3,6,9-Trioxaundecanoic Acid, Application: Dimeric RGD peptide for click chemistry, Salt Form:, Trifluoracetate salt, Storage Temperature:
Catalog No. 50-168-6959