Learn More
Medchemexpress LLC Mal-PEG3-NH2 TFA | 2830214-77-0 | 99.1% | 386.32 g/mol | C14H21F3N2O7 | 25 MG
SDP
Catalog No. 5000282214
Encompass_Preferred
Mal-PEG3-NH2 TFA is a linear heterobifunctional polyethylene glycol crosslinker supplied as the trifluoroacetic acid (TFA) salt. It features a maleimide group for selective thiol conjugation and a primary amine for subsequent modification, making it useful as a non-degradable PROTAC linker and general bioconjugation reagent for research applications.
  • Maleimide and primary amine functional groups for orthogonal conjugation.
  • Supplied as TFA salt for enhanced stability and handling.
  • High purity suitable for research-scale bioconjugation (≈99.1%).
  • Available in small mg packaging for precision experiments.
  • Non-degradable (non-cleavable) linker for stable conjugates.
Catalog No. 50-002-82214 Supplier Medchemexpress LLC Supplier No. HYW190796A25MG
Mal-PEG3-NH2 TFA is a linear heterobifunctional polyethylene glycol crosslinker supplied as the trifluoroacetic acid (TFA) salt. It features a maleimide group for selective thiol conjugation and a primary amine for subsequent modification, making it useful as a non-degradable PROTAC linker and general bioconjugation reagent for research applications.
  • Maleimide and primary amine functional groups for orthogonal conjugation.
  • Supplied as TFA salt for enhanced stability and handling.
  • High purity suitable for research-scale bioconjugation (≈99.1%).
  • Available in small mg packaging for precision experiments.
  • Non-degradable (non-cleavable) linker for stable conjugates.