Learn More
Medchemexpress LLC Mal-PEG3-NHS ester | 1537892-36-6 | 99.5% | 398.36 g/mol | C17H22N2O9 | 100 MG
SDP
Catalog No. 5000203797
Encompass_Preferred
Mal-PEG3-NHS ester is a noncleavable antibody-drug conjugate (ADC) linker containing a maleimide group and an N-hydroxysuccinimide (NHS) ester separated by a three-unit polyethylene glycol (PEG3) spacer. It enables conjugation to thiols and primary amines for bioconjugation and ADC synthesis in research applications.
  • Provides thiol and amine reactive groups for versatile bioconjugation.
  • PEG3 spacer increases aqueous solubility and reduces aggregation.
  • High purity suitable for research and conjugation workflows.
  • Available in multiple pack sizes for small- to mid-scale synthesis.
  • Store protected from light and under inert gas to maintain stability.
Catalog No. 50-002-03797 Supplier Medchemexpress LLC Supplier No. HY129526100MG
Mal-PEG3-NHS ester is a noncleavable antibody-drug conjugate (ADC) linker containing a maleimide group and an N-hydroxysuccinimide (NHS) ester separated by a three-unit polyethylene glycol (PEG3) spacer. It enables conjugation to thiols and primary amines for bioconjugation and ADC synthesis in research applications.
  • Provides thiol and amine reactive groups for versatile bioconjugation.
  • PEG3 spacer increases aqueous solubility and reduces aggregation.
  • High purity suitable for research and conjugation workflows.
  • Available in multiple pack sizes for small- to mid-scale synthesis.
  • Store protected from light and under inert gas to maintain stability.