Learn More
Medchemexpress LLC Mal-PEG3-NHS ester | 1537892-36-6 | 99.5% | 398.36 g/mol | C17H22N2O9 | 250 MG
SDP
Catalog No. 5000203955
Encompass_Preferred
Mal-PEG3-NHS ester is a heterobifunctional PEG3 linker containing a maleimide group and an N-hydroxysuccinimide (NHS) ester. It is used as a noncleavable linker for antibody-drug conjugate synthesis and for coupling thiol-bearing ligands to amine-functionalized targets.
  • Heterobifunctional maleimide-NHS PEG3 linker
  • Suitable for thiol-to-amine bioconjugation
  • Noncleavable linker for antibody-drug conjugate synthesis
  • High purity (99.5%)
  • Soluble in DMSO; for research use only
Catalog No. 50-002-03955 Supplier Medchemexpress LLC Supplier No. HY129526250MG
Mal-PEG3-NHS ester is a heterobifunctional PEG3 linker containing a maleimide group and an N-hydroxysuccinimide (NHS) ester. It is used as a noncleavable linker for antibody-drug conjugate synthesis and for coupling thiol-bearing ligands to amine-functionalized targets.
  • Heterobifunctional maleimide-NHS PEG3 linker
  • Suitable for thiol-to-amine bioconjugation
  • Noncleavable linker for antibody-drug conjugate synthesis
  • High purity (99.5%)
  • Soluble in DMSO; for research use only