Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-PEG36-NHS ester | 00-00-0 | 95.0% | C86H159N3O43 | 10MG
SDP

Catalog No. 5000259037
Encompass_Preferred

Mal-PEG36-NHS ester is a polyethylene glycol (PEG)-based bifunctional linker bearing a maleimide and an N-hydroxysuccinimide (NHS) activated ester. Supplied as a research-use solid, it facilitates thiol and amine conjugation for PROTAC assembly and other bioconjugation chemistries.

  • Enables thiol coupling via maleimide and amine coupling via NHS ester.
  • PEG36 spacer provides hydrophilicity and flexible linker length.
  • High molecular weight (≈1,923 Da) for extended linker reach.
  • Purity approximately 95.0% as provided by manufacturer.
  • Soluble in DMSO at ~62.5 mg/mL; documented in vivo formulations exist.
  • Recommended storage: -20°C for solid; low-temperature storage for solutions to preserve stability.
  • Intended for research use only; not for clinical applications.

Catalog No. 50-002-59037 Supplier Medchemexpress LLC Supplier No. HY13338910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-PEG36-NHS ester is a polyethylene glycol (PEG)-based bifunctional linker bearing a maleimide and an N-hydroxysuccinimide (NHS) activated ester. Supplied as a research-use solid, it facilitates thiol and amine conjugation for PROTAC assembly and other bioconjugation chemistries.

  • Enables thiol coupling via maleimide and amine coupling via NHS ester.
  • PEG36 spacer provides hydrophilicity and flexible linker length.
  • High molecular weight (≈1,923 Da) for extended linker reach.
  • Purity approximately 95.0% as provided by manufacturer.
  • Soluble in DMSO at ~62.5 mg/mL; documented in vivo formulations exist.
  • Recommended storage: -20°C for solid; low-temperature storage for solutions to preserve stability.
  • Intended for research use only; not for clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.