Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-PEG36-NHS ester | 95.0% | 1923.18 g/mol | C86H159N3O43 | 5 MG
SDP

Catalog No. 5000262502
Encompass_Preferred

Mal-PEG36-NHS ester is a polyethylene glycol (PEG)-based bifunctional linker bearing a maleimide and an N-hydroxysuccinimide (NHS) ester. Supplied for research use, it is designed for bioconjugation and PROTAC synthesis, enabling sequential coupling to thiol and amine groups.

  • Maleimide and NHS ester functional groups for thiol and amine coupling.
  • PEG spacer (36 units) provides flexibility and solubility.
  • Molecular weight 1923.18 g/mol.
  • Purity 95.0%.
  • Soluble in DMSO at 62.5 mg/mL (requires ultrasonic agitation).
  • Storage: pure form -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for research use only.

Catalog No. 50-002-62502 Supplier Medchemexpress LLC Supplier No. HY1333895MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-PEG36-NHS ester is a polyethylene glycol (PEG)-based bifunctional linker bearing a maleimide and an N-hydroxysuccinimide (NHS) ester. Supplied for research use, it is designed for bioconjugation and PROTAC synthesis, enabling sequential coupling to thiol and amine groups.

  • Maleimide and NHS ester functional groups for thiol and amine coupling.
  • PEG spacer (36 units) provides flexibility and solubility.
  • Molecular weight 1923.18 g/mol.
  • Purity 95.0%.
  • Soluble in DMSO at 62.5 mg/mL (requires ultrasonic agitation).
  • Storage: pure form -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.