Learn More
Medchemexpress LLC Mal-PEG4-C2-NH2 TFA | 2512227-13-1 | 430.37 | 250 MG
SDP
Catalog No. 5000351564
Encompass_Preferred
Mal-PEG4-C2-NH2 TFA is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed with two distinct ligands connected by a linker: one targets an E3 ubiquitin ligase, and the other targets a specific protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Targets E3 ubiquitin ligase and target proteins
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-51564 Supplier Medchemexpress LLC Supplier No. 250MGHY138380
Mal-PEG4-C2-NH2 TFA is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed with two distinct ligands connected by a linker: one targets an E3 ubiquitin ligase, and the other targets a specific protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Targets E3 ubiquitin ligase and target proteins
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins