Learn More
Medchemexpress LLC Mal-PEG4-C2-NH2 TFA | 2512227-13-1 | 98.6% | 430.37 | 500 MG
SDP
Catalog No. 5000350116
Encompass_Preferred
Mal-PEG4-C2-NH2 TFA is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
Catalog No. 50-003-50116 Supplier Medchemexpress LLC Supplier No. HY138380500MG
Mal-PEG4-C2-NH2 TFA is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins