Learn More
Medchemexpress LLC Mal-PEG4-NHS ester | 1325208-25-0 | 99.4% | 442.42 | 50 MG
SDP
Catalog No. 5000365990
Encompass_Preferred
Mal-PEG4-NHS ester is a non-cleavable ADC linker. It is intended for research use only and not for sale to patients. This linker connects Quantum dots (QDs) with PEGylated liposomes.
  • Non-cleavable linker
  • Suitable for laboratory research
  • High purity
  • Multiple available sizes
Catalog No. 50-003-65990 Supplier Medchemexpress LLC Supplier No. 50MGHY126505
Mal-PEG4-NHS ester is a non-cleavable ADC linker. It is intended for research use only and not for sale to patients. This linker connects Quantum dots (QDs) with PEGylated liposomes.
  • Non-cleavable linker
  • Suitable for laboratory research
  • High purity
  • Multiple available sizes