Learn More
Medchemexpress LLC Mal-PEG4-NHS ester | 1325208-25-0 | 99.4% | 442.42 | C19H26N2O10 | 250 MG
SDP
Catalog No. 5000203208
Encompass_Preferred
Mal-PEG4-NHS ester is a PEG-based crosslinker containing a maleimide group and an N-hydroxysuccinimide (NHS) ester separated by a four-unit polyethylene glycol spacer. It is used for conjugation of thiol-containing molecules to primary amines for bioconjugation and nanoparticle functionalization in research applications.
  • Non-cleavable maleimide-NHS linker for thiol-to-amine conjugation.
  • Four-unit polyethylene glycol spacer increases solubility.
  • High purity (99.4% by HPLC).
  • Molecular weight 442.42, formula C19H26N2O10.
  • Available in small lab pack sizes, including 250 MG.
  • For research use only; not for therapeutic use.
Catalog No. 50-002-03208 Supplier Medchemexpress LLC Supplier No. HY126505250MG
Mal-PEG4-NHS ester is a PEG-based crosslinker containing a maleimide group and an N-hydroxysuccinimide (NHS) ester separated by a four-unit polyethylene glycol spacer. It is used for conjugation of thiol-containing molecules to primary amines for bioconjugation and nanoparticle functionalization in research applications.
  • Non-cleavable maleimide-NHS linker for thiol-to-amine conjugation.
  • Four-unit polyethylene glycol spacer increases solubility.
  • High purity (99.4% by HPLC).
  • Molecular weight 442.42, formula C19H26N2O10.
  • Available in small lab pack sizes, including 250 MG.
  • For research use only; not for therapeutic use.