Learn More
Selleck Chemical LLC Mal-PEG4-NHS ester S0654-5MG
SDP
Catalog No. 5000042573
Encompass_Preferred
Mal-PEG4-NHS ester is a non-cleavable ADC linker containing a Maleimide group 4-unit PEG and an NHS ester
Catalog No. 50-000-42573 Supplier Selleck Chemical LLC Supplier No. S06545MG
Mal-PEG4-NHS ester is a non-cleavable ADC linker containing a Maleimide group 4-unit PEG and an NHS ester