Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-PEG4-PFP ester | 1415800-42-8 | MFCD28556926 | 95.4% | 511.39 g/mol | C21H22F5NO8 | 250 MG
SDP

Catalog No. 5000202451
Encompass_Preferred

Mal-PEG4-PFP ester is a noncleavable antibody-drug conjugate (ADC) linker composed of a maleimide group, a four-unit polyethylene glycol (PEG4) spacer, and a pentafluorophenyl (PFP) ester reactive group. It is supplied as a viscous liquid for research use only and is not intended for therapeutic use.

  • Noncleavable ADC linker suitable for conjugation strategies.
  • Maleimide group enables thiol-selective conjugation.
  • PFG ester (pentafluorophenyl) is reactive toward amines for efficient coupling.
  • PEG4 spacer increases solubility and provides distance between conjugated components.
  • High purity (95.4%) for research applications.
  • Molecular weight 511.39 g/mol and formula C21H22F5NO8.
  • Appearance viscous liquid, off-white to light brown.
  • Recommended storage: pure form -20°C for long term; in solvent store cold as specified.

Catalog No. 50-002-02451 Supplier Medchemexpress LLC Supplier No. HY126506250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-PEG4-PFP ester is a noncleavable antibody-drug conjugate (ADC) linker composed of a maleimide group, a four-unit polyethylene glycol (PEG4) spacer, and a pentafluorophenyl (PFP) ester reactive group. It is supplied as a viscous liquid for research use only and is not intended for therapeutic use.

  • Noncleavable ADC linker suitable for conjugation strategies.
  • Maleimide group enables thiol-selective conjugation.
  • PFG ester (pentafluorophenyl) is reactive toward amines for efficient coupling.
  • PEG4 spacer increases solubility and provides distance between conjugated components.
  • High purity (95.4%) for research applications.
  • Molecular weight 511.39 g/mol and formula C21H22F5NO8.
  • Appearance viscous liquid, off-white to light brown.
  • Recommended storage: pure form -20°C for long term; in solvent store cold as specified.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.