Learn More
Medchemexpress LLC Mal-PEG4-PFP ester | 1415800-42-8 | MFCD28556926 | >98.0% | 511.39 g/mol | C21H22F5NO8 | 100 MG
SDP
Catalog No. 5000202774
Encompass_Preferred
Mal-PEG4-PFP ester is a non-cleavable antibody-drug conjugate (ADC) linker containing a maleimide thiol-reactive group, a four-unit polyethylene glycol (PEG4) spacer, and a pentafluorophenyl (PFP) activated ester for bioconjugation. It is supplied as a solid for research use.
  • Non-cleavable maleimide PEG4 linker
  • PFP ester for activated carboxyl conjugation
  • Suitable for ADC and bioconjugation workflows
  • Solid powder form, typically supplied at 100 mg
  • Intended for research use only
Catalog No. 50-002-02774 Supplier Medchemexpress LLC Supplier No. HY126506100MG
Mal-PEG4-PFP ester is a non-cleavable antibody-drug conjugate (ADC) linker containing a maleimide thiol-reactive group, a four-unit polyethylene glycol (PEG4) spacer, and a pentafluorophenyl (PFP) activated ester for bioconjugation. It is supplied as a solid for research use.
  • Non-cleavable maleimide PEG4 linker
  • PFP ester for activated carboxyl conjugation
  • Suitable for ADC and bioconjugation workflows
  • Solid powder form, typically supplied at 100 mg
  • Intended for research use only