Learn More
Medchemexpress LLC Mal-PEG4-propargyl | 1262681-30-0 | 99.68% | 311.33 | 100 MG
SDP
Catalog No. 5000347980
Encompass_Preferred
Mal-PEG4-propargyl is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. For research use only.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Alkyne group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-003-47980 Supplier Medchemexpress LLC Supplier No. HY140037100MG
Mal-PEG4-propargyl is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. For research use only.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Alkyne group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups