Learn More
Medchemexpress LLC Mal-PEG4-VA-PBD | 1342820-68-1 | 1294.40 g/mol | C68H79N9O17 | 1 MG
SDP
Catalog No. 5000203248
Encompass_Preferred
Mal-PEG4-VA-PBD is an agent-linker conjugate for antibody-drug conjugate (ADC) development that incorporates a pyrrolobenzodiazepine (PBD) cytotoxic warhead linked via a maleimide-PEG4-valine-alanine (Mal-PEG4-VA) cleavable linker. It is intended for bioconjugation to antibodies through thiol-reactive maleimide chemistry.
  • Agent-linker conjugate for ADC development.
  • Pyrrolobenzodiazepine (PBD) cytotoxic payload.
  • Maleimide-PEG4-valine-alanine cleavable linker for thiol conjugation.
  • Suitable for bioconjugation to antibodies and other thiol-containing biomolecules.
  • Supplied in milligram quantities for research use.
Catalog No. 50-002-03248 Supplier Medchemexpress LLC Supplier No. HY1266851MG
Mal-PEG4-VA-PBD is an agent-linker conjugate for antibody-drug conjugate (ADC) development that incorporates a pyrrolobenzodiazepine (PBD) cytotoxic warhead linked via a maleimide-PEG4-valine-alanine (Mal-PEG4-VA) cleavable linker. It is intended for bioconjugation to antibodies through thiol-reactive maleimide chemistry.
  • Agent-linker conjugate for ADC development.
  • Pyrrolobenzodiazepine (PBD) cytotoxic payload.
  • Maleimide-PEG4-valine-alanine cleavable linker for thiol conjugation.
  • Suitable for bioconjugation to antibodies and other thiol-containing biomolecules.
  • Supplied in milligram quantities for research use.