Learn More
Medchemexpress LLC Mal-PEG4-Val-Cit-PAB | 1949793-41-2 | 98.9% | 777.86 | C36H55N7O12 | 10 MG
SDP
Catalog No. 5000204186
Encompass_Preferred
Mal-PEG4-Val-Cit-PAB is a cleavable antibody-drug conjugate (ADC) linker containing a terminal maleimide group for thiol conjugation. The construct includes a PEG4 spacer and a valine-citrulline-PAB protease-cleavable sequence to enable enzymatic payload release after internalization.
  • Cleavable valine-citrulline-PAB sequence for enzyme-triggered release.
  • Maleimide terminal for efficient thiol conjugation to antibodies.
  • PEG4 spacer increases solubility and reduces steric hindrance.
  • High supplied purity for consistent conjugation performance.
  • Soluble in DMSO; recommended storage sealed and away from moisture.
Catalog No. 50-002-04186 Supplier Medchemexpress LLC Supplier No. HY12667210MG
Mal-PEG4-Val-Cit-PAB is a cleavable antibody-drug conjugate (ADC) linker containing a terminal maleimide group for thiol conjugation. The construct includes a PEG4 spacer and a valine-citrulline-PAB protease-cleavable sequence to enable enzymatic payload release after internalization.
  • Cleavable valine-citrulline-PAB sequence for enzyme-triggered release.
  • Maleimide terminal for efficient thiol conjugation to antibodies.
  • PEG4 spacer increases solubility and reduces steric hindrance.
  • High supplied purity for consistent conjugation performance.
  • Soluble in DMSO; recommended storage sealed and away from moisture.