Learn More
Medchemexpress LLC Mal-PEG4-Val-Cit-PAB-MMAE | 00-00-0 | 98.0% | C73H115N11O19 | 10MG
SDP
Catalog No. 5000260868
Encompass_Preferred
Mal-PEG4-Val-Cit-PAB-MMAE is a drug-linker conjugate intended for antibody-drug conjugate (ADC) research. It pairs a protease-cleavable Val-Cit-PAB peptide linker with the potent MMAE payload and a maleimide thiol-reactive handle, providing a reagent for thiol-based conjugation and intracellular, enzyme-triggered payload release.
Catalog No. 50-002-60868 Supplier Medchemexpress LLC Supplier No. HY13972610MG
Mal-PEG4-Val-Cit-PAB-MMAE is a drug-linker conjugate intended for antibody-drug conjugate (ADC) research. It pairs a protease-cleavable Val-Cit-PAB peptide linker with the potent MMAE payload and a maleimide thiol-reactive handle, providing a reagent for thiol-based conjugation and intracellular, enzyme-triggered payload release.