Learn More
Medchemexpress LLC Mal-PEG4-Val-Cit-PAB-MMAE | 98.0% | 1450.76 | 25 MG
SDP
Catalog No. 5000351592
Encompass_Preferred
Mal-PEG4-Val-Cit-PAB-MMAE is a drug-linker conjugate designed for Antibody-Drug Conjugate (ADC) research. It comprises a degradable ADC linker and a potent tubulin inhibitor.
  • Contains a degradable ADC linker, Mal-PEG4-Val-Cit-PAB
  • Contains a potent tubulin inhibitor, MMAE
Catalog No. 50-003-51592 Supplier Medchemexpress LLC Supplier No. 25MGHY139726
Mal-PEG4-Val-Cit-PAB-MMAE is a drug-linker conjugate designed for Antibody-Drug Conjugate (ADC) research. It comprises a degradable ADC linker and a potent tubulin inhibitor.
  • Contains a degradable ADC linker, Mal-PEG4-Val-Cit-PAB
  • Contains a potent tubulin inhibitor, MMAE