Learn More
Medchemexpress LLC Mal-PEG4-Val-Cit-PAB-MMAE | 98.0% | 1450.76 | C73H115N11O19 | 5 MG
SDP
Catalog No. 5000223065
Encompass_Preferred
Mal-PEG4-Val-Cit-PAB-MMAE is a cleavable drug-linker conjugate used for antibody-drug conjugate research. It combines a maleimide-terminated PEG4 spacer, a Val-Cit-PAB cleavable dipeptide linker, and the tubulin inhibitor MMAE, supplied as a high-purity material for linker-payload conjugation and analytical characterization.
  • Cleavable Val-Cit-PAB linker designed for enzymatic release in lysosomes.
  • Maleimide-terminated PEG4 spacer for thiol conjugation.
  • Contains MMAE payload for potent antimitotic activity.
  • High reported purity enabling reliable conjugation and analysis.
  • Pack sizes suitable for small-scale research and method development.
Catalog No. 50-002-23065 Supplier Medchemexpress LLC Supplier No. HY1397265MG
Mal-PEG4-Val-Cit-PAB-MMAE is a cleavable drug-linker conjugate used for antibody-drug conjugate research. It combines a maleimide-terminated PEG4 spacer, a Val-Cit-PAB cleavable dipeptide linker, and the tubulin inhibitor MMAE, supplied as a high-purity material for linker-payload conjugation and analytical characterization.
  • Cleavable Val-Cit-PAB linker designed for enzymatic release in lysosomes.
  • Maleimide-terminated PEG4 spacer for thiol conjugation.
  • Contains MMAE payload for potent antimitotic activity.
  • High reported purity enabling reliable conjugation and analysis.
  • Pack sizes suitable for small-scale research and method development.