Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-PEG4-Val-Cit-PAB-PNP | 2112738-09-5 | 871.89 | C40H53N7O15 | 5 MG
SDP

Catalog No. 5000202624
Encompass_Preferred

Mal-PEG4-Val-Cit-PAB-PNP is a protease-cleavable, four-unit polyethylene glycol (PEG4) ADC linker containing a maleimide reactive group and a Val-Cit-PAB cleavable spacer. It is used in the synthesis of antibody-drug conjugates to provide protease-sensitive release of payloads.

  • Protease-cleavable PEG4 linker useful for ADC synthesis.
  • Maleimide reactive group enables thiol conjugation to antibodies.
  • Val-Cit-PAB spacer provides protease-sensitive payload release.
  • Soluble in DMSO (125 mg/mL) and formulatable for in vivo use (solubility ≥ 2.08 mg/mL with recommended solvent systems).
  • Storage: protect from light; in solvent -80°C for 6 months, -20°C for 1 month.
  • For research use only; not fully validated for medical applications.

Catalog No. 50-002-02624 Supplier Medchemexpress LLC Supplier No. HY1401455MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-PEG4-Val-Cit-PAB-PNP is a protease-cleavable, four-unit polyethylene glycol (PEG4) ADC linker containing a maleimide reactive group and a Val-Cit-PAB cleavable spacer. It is used in the synthesis of antibody-drug conjugates to provide protease-sensitive release of payloads.

  • Protease-cleavable PEG4 linker useful for ADC synthesis.
  • Maleimide reactive group enables thiol conjugation to antibodies.
  • Val-Cit-PAB spacer provides protease-sensitive payload release.
  • Soluble in DMSO (125 mg/mL) and formulatable for in vivo use (solubility ≥ 2.08 mg/mL with recommended solvent systems).
  • Storage: protect from light; in solvent -80°C for 6 months, -20°C for 1 month.
  • For research use only; not fully validated for medical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.