Learn More
Medchemexpress LLC Mal-PEG4-VC-PAB-DMEA-PNU-159682 | 2259318-52-8 | 1559.62 | C74H98N10O27 | 10 MG
SDP
Catalog No. 5000202782
Encompass_Preferred
Mal-PEG4-VC-PAB-DMEA-PNU-159682 is a cleavable antibody-drug conjugate (ADC) drug-linker conjugate that combines a Mal-PEG4-VC-PAB cleavable linker with the cytotoxic payload DMEA-PNU-159682 for use in ADC research and drug-conjugate development.
  • Cleavable ADC drug-linker conjugate suitable for antibody-drug conjugate synthesis.
  • Contains a maleimide functional group for thiol conjugation.
  • High molecular weight for payload delivery and characterization.
  • Supplied in small research pack sizes for controlled studies.
  • Store solid at -20°C, protect from light, and keep under inert gas; follow solvent storage recommendations for solutions.
Catalog No. 50-002-02782 Supplier Medchemexpress LLC Supplier No. HY12668710MG
Mal-PEG4-VC-PAB-DMEA-PNU-159682 is a cleavable antibody-drug conjugate (ADC) drug-linker conjugate that combines a Mal-PEG4-VC-PAB cleavable linker with the cytotoxic payload DMEA-PNU-159682 for use in ADC research and drug-conjugate development.
  • Cleavable ADC drug-linker conjugate suitable for antibody-drug conjugate synthesis.
  • Contains a maleimide functional group for thiol conjugation.
  • High molecular weight for payload delivery and characterization.
  • Supplied in small research pack sizes for controlled studies.
  • Store solid at -20°C, protect from light, and keep under inert gas; follow solvent storage recommendations for solutions.