Learn More
Medchemexpress LLC Mal-PEG4-VC-PAB-DMEA-PNU-159682 | 2259318-52-8 | 96.2% | 1559.62 | C74H98N10O27 | 1 MG
SDP
Catalog No. 5000203298
Encompass_Preferred
Mal-PEG4-VC-PAB-DMEA-PNU-159682 is a drug-linker conjugate used in antibody-drug conjugate (ADC) research. It combines a cleavable maleimide-PEG4-valine-citrulline-PAB-DMEA linker with a potent cytotoxic payload to enable studies of conjugation chemistry, linker stability, and payload release in vitro.
  • Cleavable valine-citrulline peptide linker enabling enzymatic release.
  • Maleimide functional group for thiol conjugation to antibodies.
  • PEG4 spacer to improve solubility and spacing between antibody and payload.
  • Conjugated to a potent cytotoxin for payload delivery studies.
  • Supplied in small pack sizes suitable for early stage research.
Catalog No. 50-002-03298 Supplier Medchemexpress LLC Supplier No. HY1266871MG
Mal-PEG4-VC-PAB-DMEA-PNU-159682 is a drug-linker conjugate used in antibody-drug conjugate (ADC) research. It combines a cleavable maleimide-PEG4-valine-citrulline-PAB-DMEA linker with a potent cytotoxic payload to enable studies of conjugation chemistry, linker stability, and payload release in vitro.
  • Cleavable valine-citrulline peptide linker enabling enzymatic release.
  • Maleimide functional group for thiol conjugation to antibodies.
  • PEG4 spacer to improve solubility and spacing between antibody and payload.
  • Conjugated to a potent cytotoxin for payload delivery studies.
  • Supplied in small pack sizes suitable for early stage research.