Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-PEG5-acid | 1286755-26-7 | ≥97.0% | 25 MG
SDP

Catalog No. 5000338896
Encompass_Preferred

Mal-PEG5-acid is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. These molecules contain two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein. PROTACs function by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is intended for research use only.

  • PEG-based PROTAC linker for chemical synthesis
  • Utilizes the ubiquitin-proteasome system for selective protein degradation
  • Molecular weight: 389.40
  • Chemical formula: C17H27NO9
  • Purity: ≥97.0%
  • Appearance: Light brown to yellow liquid
  • Soluble in DMSO (100 mg/mL)

Catalog No. 50-003-38896 Supplier Medchemexpress LLC Supplier No. 25MGHY126962
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-PEG5-acid is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. These molecules contain two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein. PROTACs function by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is intended for research use only.

  • PEG-based PROTAC linker for chemical synthesis
  • Utilizes the ubiquitin-proteasome system for selective protein degradation
  • Molecular weight: 389.40
  • Chemical formula: C17H27NO9
  • Purity: ≥97.0%
  • Appearance: Light brown to yellow liquid
  • Soluble in DMSO (100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.