Learn More
Medchemexpress LLC Mal-PEG5-NHS ester | 1807537-42-3 | 99.0% | 10 MG
SDP
Catalog No. 5000341330
Encompass_Preferred
Mal-PEG5-NHS ester is an Alkyl/ether and PEG-based PROTAC linker. It is used in the synthesis of PROTACs, which are composed of two different ligands connected by a linker. One ligand is for an E3 ubiquitin ligase and the other is for the target protein. PROTACs function by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl/ether and PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-41330 Supplier Medchemexpress LLC Supplier No. HY12688810MG
Mal-PEG5-NHS ester is an Alkyl/ether and PEG-based PROTAC linker. It is used in the synthesis of PROTACs, which are composed of two different ligands connected by a linker. One ligand is for an E3 ubiquitin ligase and the other is for the target protein. PROTACs function by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl/ether and PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins